N-[4-(sec-Butoxy)phenyl]-N-[2-(4-methylphenoxy)butyl]amine

Supplier: Matrix Scientific

031321-500MG
101831-468EA 306.15 USD
101831-468
N-[4-(sec-Butoxy)phenyl]-N-[2-(4-methylphenoxy)butyl]amine
N-[4-(sec-Butoxy)phenyl]-N-[2-(4-methylphenoxy)butyl]amine

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MW: 327.47 g/mol
MDL Number: MFCD10687452
CAS Number: 1040685-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR