N-[4-(sec-Butoxy)phenyl]-N-[2-(2,5-dimethylphenoxy)propyl]amine

Supplier: Matrix Scientific

031320-500MG
101831-466EA 306.15 USD
101831-466
N-[4-(sec-Butoxy)phenyl]-N-[2-(2,5-dimethylphenoxy)propyl]amine
N-[4-(sec-Butoxy)phenyl]-N-[2-(2,5-dimethylphenoxy)propyl]amine

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10687451
CAS Number: 1040685-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR