4-(sec-Butoxy)-N-(2-phenoxypropyl)aniline

Supplier: Matrix Scientific

031319-500MG
101831-464EA 515.38 USD
101831-464
4-(sec-Butoxy)-N-(2-phenoxypropyl)aniline
4-(sec-Butoxy)-N-(2-phenoxypropyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687450
CAS Number: 1040685-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR