N-[4-(sec-Butoxy)phenyl]-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]amine

Supplier: Matrix Scientific

031316-500MG
101831-458EA 306.15 USD
101831-458
N-[4-(sec-Butoxy)phenyl]-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]amine
N-[4-(sec-Butoxy)phenyl]-N-[2-(2-isopropyl-5-methylphenoxy)ethyl]amine

Hazard: Irritant

Formula: C₂₂H₃₁NO₂
MDL Number: MFCD10687447
CAS Number: 1040684-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR