Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[4-(sec-Butoxy)benzyl]-3-isopropoxyaniline

Supplier: Matrix Scientific


031314-500MG
101831-454EA 260.23 USD InStock
101831-454
N-[4-(sec-Butoxy)benzyl]-3-isopropoxyaniline
N-[4-(sec-Butoxy)benzyl]-3-isopropoxyaniline
CAS: 1040684-82-9
MDL Number: MFCD10687445
Molecular Formula: C20H27NO2
Molecular Weight: 313,44
Hazard: Irritant


Formula: C₂₀H₂₇NO₂
MW: 313,44 g/mol
MDL Number: MFCD10687445
CAS Number: 1040684-82-9
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty