N-[4-(2-Ethoxyethoxy)benzyl]-3-isopropoxyaniline
Supplier: Matrix Scientific
031313-500MG
101831-452EA
306.15
USD
101831-452
N-[4-(2-Ethoxyethoxy)benzyl]-3-isopropoxyaniline
N-[4-(2-Ethoxyethoxy)benzyl]-3-isopropoxyaniline
Hazard: Irritant
Formula:
C₂₀H₂₇NO₃ |
MDL Number:
MFCD10687444 CAS Number: 1040684-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...