3-Isopropoxy-N-[4-(phenethyloxy)benzyl]aniline

Supplier: Matrix Scientific

031312-500MG
101831-450EA 306.15 USD
101831-450
3-Isopropoxy-N-[4-(phenethyloxy)benzyl]aniline
3-Isopropoxy-N-[4-(phenethyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₄H₂₇NO₂
MDL Number: MFCD10687443
CAS Number: 1040684-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR