3-Isopropoxy-N-(4-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

031311-500MG
101831-448EA 306.15 USD
101831-448
3-Isopropoxy-N-(4-isopropoxybenzyl)aniline
3-Isopropoxy-N-(4-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687442
CAS Number: 1040684-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR