N-(2-Isobutoxybenzyl)-3-isopropoxyaniline

Supplier: Matrix Scientific

031309-500MG
101831-444EA 306.15 USD
101831-444
N-(2-Isobutoxybenzyl)-3-isopropoxyaniline
N-(2-Isobutoxybenzyl)-3-isopropoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10687440
CAS Number: 1040684-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR