3-Isopropoxy-N-[2-(2-methoxyphenoxy)propyl]aniline

Supplier: Matrix Scientific

031307-500MG
101831-440EA 306.15 USD
101831-440
3-Isopropoxy-N-[2-(2-methoxyphenoxy)propyl]aniline
3-Isopropoxy-N-[2-(2-methoxyphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₃
MDL Number: MFCD10687438
CAS Number: 1040684-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR