3-Isopropoxy-N-(4-methoxybenzyl)aniline

Supplier: Matrix Scientific

031304-500MG
101831-434EA 306.15 USD
101831-434
3-Isopropoxy-N-(4-methoxybenzyl)aniline
3-Isopropoxy-N-(4-methoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁NO₂
MDL Number: MFCD10687435
CAS Number: 1040684-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR