3-Isopropoxy-N-[2-(4-isopropylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031303-500MG
101831-432EA 306.15 USD
101831-432
3-Isopropoxy-N-[2-(4-isopropylphenoxy)ethyl]aniline
3-Isopropoxy-N-[2-(4-isopropylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MW: 313.44 g/mol
MDL Number: MFCD10687434
CAS Number: 1040684-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR