2-Isopropoxy-N-(3-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

031301-500MG
101831-428EA 306.15 USD
101831-428
2-Isopropoxy-N-(3-isopropoxybenzyl)aniline
2-Isopropoxy-N-(3-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687432
CAS Number: 1040683-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR