N-[2-(2,6-Dimethylphenoxy)propyl]-2-isopropoxyaniline

Supplier: Matrix Scientific

031297-500MG
101831-420EA 306.15 USD
101831-420
N-[2-(2,6-Dimethylphenoxy)propyl]-2-isopropoxyaniline
N-[2-(2,6-Dimethylphenoxy)propyl]-2-isopropoxyaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10687428
CAS Number: 1040683-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR