2-Isopropoxy-N-[2-(4-methylphenoxy)butyl]aniline

Supplier: Matrix Scientific

031292-500MG
101831-410EA 306.15 USD
101831-410
2-Isopropoxy-N-[2-(4-methylphenoxy)butyl]aniline
2-Isopropoxy-N-[2-(4-methylphenoxy)butyl]aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10687423
CAS Number: 1040683-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR