2-Isopropoxy-N-(4-methoxyphenethyl)aniline

Supplier: Matrix Scientific

031288-500MG
101831-402EA 306.15 USD
101831-402
2-Isopropoxy-N-(4-methoxyphenethyl)aniline
2-Isopropoxy-N-(4-methoxyphenethyl)aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10687419
CAS Number: 1040682-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR