N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2-isopropoxyaniline

Supplier: Matrix Scientific

031285-500MG
101831-396EA 306.15 USD
101831-396
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2-isopropoxyaniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2-isopropoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₄ClNO₂
MDL Number: MFCD10687416
CAS Number: 1040682-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR