3-(Phenethyloxy)-N-[2-(phenethyloxy)benzyl]aniline

Supplier: Matrix Scientific

031283-500MG
101831-392EA 306.15 USD
101831-392
3-(Phenethyloxy)-N-[2-(phenethyloxy)benzyl]aniline
3-(Phenethyloxy)-N-[2-(phenethyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₉H₂₉NO₂
MDL Number: MFCD10687414
CAS Number: 1040682-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR