N-[2-(2,6-Dimethylphenoxy)propyl]-3-(phenethyloxy)aniline

Supplier: Matrix Scientific

031281-500MG
101831-388EA 306.15 USD
101831-388
N-[2-(2,6-Dimethylphenoxy)propyl]-3-(phenethyloxy)aniline
N-[2-(2,6-Dimethylphenoxy)propyl]-3-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₅H₂₉NO₂
MDL Number: MFCD10687412
CAS Number: 1040682-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR