3-(Phenethyloxy)-N-[4-(phenethyloxy)benzyl]aniline

Supplier: Matrix Scientific

031280-500MG
101831-386EA 306.15 USD
101831-386
3-(Phenethyloxy)-N-[4-(phenethyloxy)benzyl]aniline
3-(Phenethyloxy)-N-[4-(phenethyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₉H₂₉NO₂
MDL Number: MFCD10687411
CAS Number: 1040682-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR