N-[4-(2,4-Dichlorophenoxy)butyl]-3-(phenethyloxy)aniline

Supplier: Matrix Scientific

031275-500MG
101831-376EA 306.15 USD
101831-376
N-[4-(2,4-Dichlorophenoxy)butyl]-3-(phenethyloxy)aniline
N-[4-(2,4-Dichlorophenoxy)butyl]-3-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₂₄H₂₅Cl₂NO₂
MDL Number: MFCD10687406
CAS Number: 1040681-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR