3-(Phenethyloxy)-N-(3-phenylpropyl)aniline

Supplier: Matrix Scientific

031274-500MG
101831-374EA 306.15 USD
101831-374
3-(Phenethyloxy)-N-(3-phenylpropyl)aniline
3-(Phenethyloxy)-N-(3-phenylpropyl)aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO
MDL Number: MFCD10687405
CAS Number: 1040681-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR