N-[4-(2-Ethoxyethoxy)benzyl]-1-naphthalenamine

Supplier: Matrix Scientific

031273-500MG
101831-372EA 306.15 USD
101831-372
N-[4-(2-Ethoxyethoxy)benzyl]-1-naphthalenamine
N-[4-(2-Ethoxyethoxy)benzyl]-1-naphthalenamine

Hazard: Irritant

Formula: C₂₁H₂₃NO₂
MDL Number: MFCD10687404
CAS Number: 1040681-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR