2-Fluoro-4-(trifluoromethyl)nicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid

031264-500MG
101831-346EA 113.85 USD
101831-346
2-Fluoro-4-(trifluoromethyl)nicotinic acid ≥97%
2-Fluoro-4-(trifluoromethyl)nicotinic acid

Hazard: Irritant

Formula: C₇H₃F₄NO₂
MW: 209.10 g/mol
MDL Number: MFCD10687403
CAS Number: 1040681-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR