6-Bromopicolinamide ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Bromopyridine-2-carboxamide

031263-1G
101831-344EA 60 USD
101831-344
6-Bromopicolinamide ≥97%
6-Bromopicolinamide

Hazard: Irritant

Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
MDL Number: MFCD00234154
CAS Number: 25194-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR