Tetrazolo[1,5-a]pyridine-6-carboxylic acid

Supplier: Matrix Scientific

031250-500MG
101831-310EA 444.62 USD
101831-310
Tetrazolo[1,5-a]pyridine-6-carboxylic acid
Tetrazolo[1,5-a]pyridine-6-carboxylic acid

Hazard: Irritant

Formula: C₆H₄N₄O₂
MDL Number: MFCD09971234
CAS Number: 7477-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR