1,3-Benzothiazole-2,4-diamine

Supplier: Matrix Scientific

031232-500MG
101831-304EA 461.54 USD
101831-304
1,3-Benzothiazole-2,4-diamine
1,3-Benzothiazole-2,4-diamine

Hazard: Irritant

Formula: C₇H₇N₃S
MDL Number: MFCD09972079
CAS Number: 861100-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR