5-(6-Chloropyridazin-3-yl)thiophene-2-sulfonyl chloride

Supplier: Matrix Scientific

031206-500MG
101831-296EA 476.92 USD
101831-296
5-(6-Chloropyridazin-3-yl)thiophene-2-sulfonyl chloride
5-(6-Chloropyridazin-3-yl)thiophene-2-sulfonyl chloride

Hazard: Irritant

Formula: C₈H₄Cl₂N₂O₂S₂
MDL Number: MFCD09972067
CAS Number: 1017791-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR