3-(2-Chlorophenyl)propan-1-amine

Supplier: Matrix Scientific

031153-500MG
101831-272EA 300 USD
101831-272
3-(2-Chlorophenyl)propan-1-amine
3-(2-Chlorophenyl)propan-1-amine

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD06212636
CAS Number: 18655-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR