3-(3-Methoxyphenyl)propan-1-amine

Supplier: Matrix Scientific

031152-500MG
101831-270EA 461.54 USD
101831-270
3-(3-Methoxyphenyl)propan-1-amine
3-(3-Methoxyphenyl)propan-1-amine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD00870509
CAS Number: 18655-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR