3-(4-Methoxyphenyl)propan-1-amine

Supplier: Matrix Scientific

031151-500MG
101831-268EA 429.23 USD
101831-268
3-(4-Methoxyphenyl)propan-1-amine
3-(4-Methoxyphenyl)propan-1-amine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
MDL Number: MFCD00870510
CAS Number: 36397-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR