(4-Bromo-1H-pyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-Bromo-1H-pyrazol-1-yl)acetic acid

031147-500MG
101831-264EA 250.77 USD
101831-264
(4-Bromo-1H-pyrazol-1-yl)acetic acid
(4-Bromo-1H-pyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₅H₅BrN₂O₂
MW: 205.0100 g/mol
MDL Number: MFCD00159632
CAS Number: 82231-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR