3-[(2-Fluorophenoxy)methyl]-4-methoxybenzaldehyde

Supplier: Matrix Scientific

031115-500MG
101831-252EA 320 USD
101831-252
3-[(2-Fluorophenoxy)methyl]-4-methoxybenzaldehyde
3-[(2-Fluorophenoxy)methyl]-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃FO₃
MDL Number: MFCD02734130
CAS Number: 351366-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR