2-Chloro-5-(4H-1,2,4-triazol-4-yl)aniline

Supplier: Matrix Scientific

031102-500MG
101831-246EA 444.62 USD
101831-246
2-Chloro-5-(4H-1,2,4-triazol-4-yl)aniline
2-Chloro-5-(4H-1,2,4-triazol-4-yl)aniline

Hazard: Irritant

Formula: C₈H₇ClN₄
MDL Number: MFCD09972035
CAS Number: 1017791-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR