2-Amino-7,7-dimethyl-7,8-dihydroquinazolin-5(6H)-one

Supplier: Matrix Scientific

031046-1G
101831-238EA 235.38 USD
101831-238
2-Amino-7,7-dimethyl-7,8-dihydroquinazolin-5(6H)-one
2-Amino-7,7-dimethyl-7,8-dihydroquinazolin-5(6H)-one

Hazard: Irritant

Formula: C₁₀H₁₃N₃O
MDL Number: MFCD00520824
CAS Number: 21599-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR