Methyl-6,7-dimethoxy-1,2,3,4-tetra-hydroisoquinoline-3-carboxylate hydrochloride

Supplier: Matrix Scientific

031044-500MG
101831-236EA 300 USD
101831-236
Methyl-6,7-dimethoxy-1,2,3,4-tetra-hydroisoquinoline-3-carboxylate hydrochloride
Methyl-6,7-dimethoxy-1,2,3,4-tetra-hydroisoquinoline-3-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₄
MDL Number: MFCD08594063
CAS Number: 30740-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR