2-Chloro-N-2,3-dihydro-1H-inden-2-ylacetamide

Supplier: Matrix Scientific

031043-500MG
101831-234EA 364.62 USD
101831-234
2-Chloro-N-2,3-dihydro-1H-inden-2-ylacetamide
2-Chloro-N-2,3-dihydro-1H-inden-2-ylacetamide

Hazard: Irritant

Formula: C₁₁H₁₂ClNO
MDL Number: MFCD01764041
CAS Number: 36851-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR