1-(Thien-2-ylcarbonyl)piperidin-4-amine
Supplier: Matrix Scientific
031022-500MG
101831-218EA
461.54
USD
101831-218
1-(Thien-2-ylcarbonyl)piperidin-4-amine
1-(Thien-2-ylcarbonyl)piperidin-4-amine
Hazard: Irritant
Formula:
C₁₀H₁₄N₂OS |
MDL Number:
MFCD06740654 CAS Number: 886497-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...