2-[(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoic acid

Supplier: Matrix Scientific

030970-500MG
101831-204EA 193.85 USD
101831-204
2-[(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoic acid
2-[(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₄
MDL Number: MFCD03466731
CAS Number: 512813-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR