3-(1-Adamantyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

030969-500MG
101831-202EA 193.85 USD
101831-202
3-(1-Adamantyl)-1-phenyl-1H-pyrazole-4-carboxylic acid
3-(1-Adamantyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₂N₂O₂
MDL Number: MFCD02090831
CAS Number: 401632-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR