2-(1H-Pyrazol-4-yl)ethanol

Supplier: Matrix Scientific

Synonyms: 4-(2-Hydroxyethyl)-1H-pyrazole

030963-500MG
101831-190EA 40 USD
101831-190
2-(1H-Pyrazol-4-yl)ethanol
2-(1H-Pyrazol-4-yl)ethanol

Hazard: Irritant

Formula: C₅H₈N₂O
MW: 112.1298 g/mol
MDL Number: MFCD03419343
CAS Number: 180207-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR