4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-2-hydrazino-6-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

030957-500MG
101831-178EA 290.77 USD
101831-178
4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-2-hydrazino-6-(trifluoromethyl)pyrimidine
4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-2-hydrazino-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₁H₁₃F₃N₆
MDL Number: MFCD04970350
CAS Number: 925580-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR