4-(4-Chlorophenyl)-6-(difluoromethyl)-2-(ethylsulfonyl)pyrimidine

Supplier: Matrix Scientific

030955-500MG
101831-174EA 193.85 USD
101831-174
4-(4-Chlorophenyl)-6-(difluoromethyl)-2-(ethylsulfonyl)pyrimidine
4-(4-Chlorophenyl)-6-(difluoromethyl)-2-(ethylsulfonyl)pyrimidine

Hazard: Irritant

Formula: C₁₃H₁₁ClF₂N₂O₂S
MDL Number: MFCD05861689
CAS Number: 862678-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR