4-(4-Ethylphenyl)-2-hydrazino-6-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

030953-500MG
101831-170EA 290.77 USD
101831-170
4-(4-Ethylphenyl)-2-hydrazino-6-(trifluoromethyl)pyrimidine
4-(4-Ethylphenyl)-2-hydrazino-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₃H₁₃F₃N₄
MDL Number: MFCD05861706
CAS Number: 861439-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR