2-Hydrazino-4-methyl-6-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

030952-500MG
101831-168EA 290.77 USD
101831-168
2-Hydrazino-4-methyl-6-(trifluoromethyl)pyrimidine
2-Hydrazino-4-methyl-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₆H₇F₃N₄
MDL Number: MFCD05861708
CAS Number: 123024-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR