4-(3,4-Dichlorophenyl)-2-hydrazino-6-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

030950-500MG
101831-164EA 290.77 USD
101831-164
4-(3,4-Dichlorophenyl)-2-hydrazino-6-(trifluoromethyl)pyrimidine
4-(3,4-Dichlorophenyl)-2-hydrazino-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₁H₇Cl₂F₃N₄
MDL Number: MFCD05861712
CAS Number: 862658-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR