4-Nitro-7-piperidin-1-yl-2,1,3-benzoxadiazole

Supplier: Matrix Scientific

030948-500MG
101831-160EA 146.15 USD
101831-160
4-Nitro-7-piperidin-1-yl-2,1,3-benzoxadiazole
4-Nitro-7-piperidin-1-yl-2,1,3-benzoxadiazole

Hazard: Irritant

Formula: C₁₁H₁₂N₄O₃
MDL Number: MFCD00576861
CAS Number: 18378-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR