2-Hydrazino-4-(2-methoxyphenyl)-6-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

030946-500MG
101831-156EA 290.77 USD
101831-156
2-Hydrazino-4-(2-methoxyphenyl)-6-(trifluoromethyl)pyrimidine
2-Hydrazino-4-(2-methoxyphenyl)-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₂H₁₁F₃N₄O
MDL Number: MFCD05861872
CAS Number: 861440-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR