7-Azepan-1-yl-2,1,3-benzoxadiazol-4-amine

Supplier: Matrix Scientific

030943-500MG
101831-150EA 290.77 USD
101831-150
7-Azepan-1-yl-2,1,3-benzoxadiazol-4-amine
7-Azepan-1-yl-2,1,3-benzoxadiazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₆N₄O
MDL Number: MFCD09431371
CAS Number: 925154-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR