2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine

Supplier: Matrix Scientific

030938-500MG
101831-140EA 541.54 USD
101831-140
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine

Hazard: Irritant

Formula: C₇H₁₀F₃N₃
MDL Number: MFCD04967724
CAS Number: 925154-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR